Tisk…
Cell culture inserts for multiwell plates, ThinCert™
Dodavatel: Greiner Bio-One
665641_48 657610_24 657630_24 657631_24 657638_24 657640_24 657641_24 662610_48 662630_48 662631_48 662638_48 662640_48 662641_48 665610_48 665630_48 665631_48 665638_48 665640_48
392-0055EA 11130 CZK
392-0055 392-0128 392-0129 392-0130 392-0131 392-0037 392-0132 392-0040 392-0041 392-0042 392-0133 392-0043 392-0044 392-0050 392-0051 392-0052 392-0053 392-0054
Cell culture inserts for multiwell plates, ThinCert™
Mikrodestičky Destičky inzertní a inzerty
Clear PS housing with PET capillary pore membranes, cell culture surface treated, sterile. Combining six different membrane specifications (pore size and density) in geometries to fit 6,12 and 24 well plates, the ThinCert™ cell culture inserts are suitable for a wide range of applications including transportation, secretion and diffusion studies, migrational experiments, cytotoxicity testing, co cultures, Trans Epithelial Electric Resistance (TEER) measurements, as well as primary cell culture.
  • Physical surface treatment optimises cell adherence and growth
  • Hanging geometry ensures
More Product Information