Tisk…

Potassium-2-ethylhexanoate solution ≥75% (w/w)

Dodavatel: Thermo Fisher Scientific
Varování

39163.18 39163.04
39163.18EA 15480 CZK
39163.18 39163.04
Potassium-2-ethylhexanoate solution ≥75% (w/w)
Potassium-2-ethylhexanoate

Purity: 99.9% CAS No.: 3164-85-0 Molecular Formula: C8H15KO2 Formula Weight: 182.30 Physical Form: Viscous liquid Odour: Characteristic UN No.: UN1993 MDL No.: MFCD00045896

Vzorec: C₈H₁₆O₂
Storage Temperature: Ambient
MDL Number: MFCD00045896
CAS číslo: 3164-85-0
EINECS: 221-625-7
UN: 1993
ADR: 3,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR