Tisk…
Bottles, narrow neck, octagonal, PET, with screw caps, Corning®
Dodavatel: Corning
431731 431734 431732 431733 431729 431730
391-0432EA 6170 CZK
391-0432 391-0435 391-0433 391-0434 391-0430 391-0431
Bottles, narrow neck, octagonal, PET, with screw caps, Corning®
Láhve Láhve plastové
PET, clear, with natural HDPE screw cap.
  • Packaged in convenient, shrinkwrapped trays within an outer bag to assure cleanliness
  • Bottles are sterilised to 10⁻⁶ SAL and certified nonpyrogenic
  • USP class VI compliant bottles are validated noncytotoxic and nonhemolytic
Ideal for storage and sampling of media, biologicals, and other aqueous solutions. The PET resin makes the bottle lightweight, break resistant and as transparent as glass. Leakproof, tamper-evident HDPE screw cap. Molded graduations for accurate measurements.