Tisk…
Culture bottles and culture flasks, DURAN®
Dodavatel: DWK Life Sciences
214212402 214213201 217712401 217713209 217713603 217714402 217715407 217716309 217713903
391-0210EA 1070 CZK
391-0210 391-0215 391-0260 391-0265 391-0270 391-0280 391-0285 391-0290 391-0275
Culture bottles and culture flasks, DURAN®
Baňky Baňky kultivační
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Culture bottles and culture flasks (erlenmeyer shape) with straight rim, suitable for either kapsenberg caps or metal caps (available separately).