Tisk…
VWR® INCU-Line®, Cooled Incubators
Dodavatel: VWR Collection
390-1341EA 73390 CZK
390-1341 390-1342
VWR® INCU-Line®, Cooled Incubators
Inkubátory Inkubátory chlazené
These compressor-cooled incubators with forced air convection provide a temperature range between 3 to 70 °C. Featuring a stainless steel (DIN 1.4016) inner chamber and powder coated sheet exterior. These models have an intuitive menu, USB interface for data download to USB stick, and can be connected to LAN network. The IL 68R has an external glass door, the IL 150R/250R models have a solid outer door and internal glass door.
  • Multi-functional microprocessor controller with 4,3" full colour touch screen
  • Internal memory stores up to 10000 data records (
More Product Information