Tisk…
Incubators with dual convection, Heratherm®, Advanced Protocol Security
Dodavatel: Thermo Fisher Scientific
51028541 51028542 51028136 51028543 51028138 51028137
390-0479EA 110500 CZK
390-0479 390-0480 390-0473 390-0481 390-0475 390-0474
Incubators with dual convection, Heratherm®, Advanced Protocol Security
Inkubátory Inkubátory standardní
Heratherm® Advanced Protocol Security incubators feature adjustable forced convection which allows the user to select the fan speed from 0 up to 100%. Air speed can be adjusted to give optimum airflow depending on the application. These units are recommended for a wide range of demanding laboratory applications as well as food and beverage and pharmaceutical stability testing. The incubators have a smooth stainless steel inner chamber with easy to clean, rounded corners. Units feature an internal glass door, allowing samples to be viewed without impacting on temperature stability, a lockableMore Product Information