Tisk…

(+)-B-Chlorodiisopinocampheylborane 1.8 M in hexanes, AcroSeal®

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: (+)-Diisopinocampheylchloroborane

388421000
388421000.EA 8640 CZK
388421000.
(+)-B-Chlorodiisopinocampheylborane 1.8 M in hexanes, AcroSeal®
(+)-B-Chlorodiisopinocampheylborane
Vzorec: C₂₀H₃₄BCl
MW: 320.75 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00074808
CAS číslo: 112246-73-8
UN: 2920
ADR: 8 I

Objednat

Výsledky specifikačního testu

Appearance Clear colorless to light yellow Solution
Titration Argentometric 1.65 to 1.85 M

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR