Tisk…

Phenol red sodium salt 0,02% (w/v) vodný roztok

Dodavatel: Thermo Fisher Scientific

Synonyms: 3,3-Bis(4-hydroxyphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide, sodium salt, Phenol red water soluble, Sodium hydrogen 4,4'-(3H-2,1-benzoxathiol-3-ylidene)bisphenolate S,S-dioxide

38702.AE 38702.AP
38702.AEEA 1050 CZK
38702.AE 38702.AP
Phenol red sodium salt 0,02% (w/v) vodný roztok
Phenol red sodium salt

Molecular Formula: C19H13NaO5S Formula Weight: 376.37 Physical Form: Liquid Density: 0.972 MDL No.: MFCD00066901

Vzorec: C₁₉H₁₃NaO₅S
MW: 376.35
Bod tání: 285
Hustota: 0.972
Storage Temperature: Ambient
MDL Number: MFCD00066901
CAS číslo: 34487-61-1
EINECS: 252-057-8
Merck Index: 12,07397

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR