Tisk…

Kyselina chlorovodíková concentrate for dilution to 1L, 0.2 M HCl (0.2N) volumetric solution, Fluka™

Dodavatel: Honeywell Chemicals
Nebezpečí

38287-1EA 38287-6X1EA
38287-1EAEA 1800 CZK
38287-1EA 38287-6X1EA
Kyselina chlorovodíková concentrate for dilution to 1L, 0.2 M HCl (0.2N) volumetric solution, Fluka™
Kyselina chlorovodíková

Concentrate for 1L standard solution

Vzorec: HCl
Hustota: 1,11 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS číslo: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8,II

Objednat

Výsledky specifikačního testu

Factor 0.998 - 1.002
BAM/EMPA reference material effective
BAM/EMPA Certificate Number effective
Traceable to NIST SRM effective


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR