Tisk…

Hydrogenfosforečnan dvojsodný heptahydrát 98-100.0% (dried basis), crystals USP, J.T.Baker®

Dodavatel: Avantor
Varování

Synonyms: Sodium phosphate dibasic heptahydrate, Sodium hydrogen phosphate heptahydrate, Sodium monohydrogen phosphate heptahydrate

3817-01EA 3350 CZK
3817-01 3817-05 3817-07
Hydrogenfosforečnan dvojsodný heptahydrát 98-100.0% (dried basis), crystals USP, J.T.Baker®
Hydrogenfosforečnan dvojsodný heptahydrát
Vzorec: HNa₂O₄P·7H₂O
Bod tání: 48 °C
Hustota: 1,7 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149180
CAS číslo: 7782-85-6
EINECS: 231-448-7

Objednat

Výsledky specifikačního testu

GMP Manufactured Product 98.0 - 100.5%
Meets U.S.P Requirements ≤16 ppm
CAUTION: For Manufacturing, processing or repackaging ≤0.06%
Bulk Pharmaceutical Chemical Passes Test
USP - Assay (Na₂HPO₄) (dried basis) 98.0 - 100.5 %
USP - Arsenic (As) ≤16 ppm
USP - Chloride (Cl) ≤0.06 %
USP - Identification Passes Test
USP - Insoluble Substances ≤0.4 %
USP - Loss on Drying at 130°C 43.0 - 50.0 %
USP - Sulfate (SO₄) ≤0.2 %
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR