Tisk…

Glassy carbon, plate, Thickness: 4 mm (0.16 in) Type 2

Dodavatel: Thermo Fisher Scientific

38023.FI 38023.GH 38023.FF
38023.FIEA 13830 CZK
38023.FI 38023.GH 38023.FF
Glassy carbon, plate, Thickness: 4 mm (0.16 in) Type 2
Glassy carbon
Vzorec: C
Bod tání: 3652 °C
Hustota: 1,95 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00133992
CAS číslo: 7440-44-0
EINECS: 231-153-3
Merck Index: 13,01818

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR