Tisk…

Glassy carbon, plate, Thickness: 1 mm (0.04 in) Type2

Dodavatel: Thermo Fisher Scientific

38021.FIEA 15070 CZK
38021.FI 38021.GH
Glassy carbon, plate, Thickness: 1 mm (0.04 in) Type2
Glassy carbon
Vzorec: C
Bod tání: 3652 °C
Hustota: 1,95 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00133992
CAS číslo: 7440-44-0
EINECS: 231-153-3
Merck Index: 13,01818

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR