Tisk…

Močovina 99,0-100,5 ACS

Dodavatel: Thermo Fisher Scientific

Synonyms: Carbonyl diamide, Carbamide, Diaminomethanone, Carbonyldiamide

36428.22 36428.36 36428.A3
36428.22EA 1230 CZK
36428.22 36428.36 36428.A3
Močovina 99,0-100,5 ACS
Močovina

Purity: 99.0-100.5% CAS No.: 57-13-6 Molecular Formula: NH2CONH2 Formula Weight: 60.06 Physical Form: Crystalline Density: 1.335 pH: 9,2-9,5 Melting Point: 132-135° Solubility: Soluble in water, alcohol, methanol, glycerol, benzene, and conc. HCl. Practically insoluble in chloroform, ether Beilstein Reference: 635724 Merck Reference: 14,9867 MDL No.: MFCD00008022


Used as a protein denaturant

Vzorec: NH₂CONH₂
MW: 60.06
Bod varu: 197
Bod tání: 132…135 °C
Hustota: 1.335
Storage Temperature: Ambient
MDL Number: MFCD00008022
CAS číslo: 57-13-6
EINECS: 200-315-5
REACH: 01-2119463277-33
Merck Index: 14,9867
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a9682f71b212a449398841e2beace2d7500b4710&vid=d6057a7ec3d606031d338a300b5d5db30b52d861&prid=web&clid=SAPDAM

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR