Tisk…

Lactic acid ≥85.0-90.0% ACS

Dodavatel: Thermo Fisher Scientific
Nebezpečí

36415.22 36415.36 36415.A3
36415.22EA 1110 CZK
36415.22 36415.36 36415.A3
Lactic acid ≥85.0-90.0% ACS
Lactic acid

CAS No.: 50-21-5 Molecular Formula: C3H6O3 Formula Weight: 90.08 Flash Point: 110°(230°F) Physical Form: Liquid Density: 1.209 Refractive Index: 1.4270 Appearance: Colourless to yellow Melting Point: 18° Boiling Point: 122°/15mm Beilstein Reference: 1209341 Merck Reference: 14,5336 MDL No.: MFCD00004520

Vzorec: H₃CCH(OH)COOH
Bod varu: 122 °C (15 mmHg)
Bod tání: 18 °C
Hustota: 1,209
Bod vzplanutí: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004520
CAS číslo: 50-21-5
EINECS: 200-018-0
Merck Index: 14,5336

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR