Tisk…

Síran nikelnatý hexahydrát ≥98.0% ACS

Dodavatel: Thermo Fisher Scientific
Nebezpečí

36336.22 36336.36 36336.A4
36336.22EA 1330 CZK
36336.22 36336.36 36336.A4
Síran nikelnatý hexahydrát ≥98.0% ACS
Síran nikelnatý hexahydrát

Purity: 98.0% min CAS No.: 10101-97-0 Molecular Formula: NiO4S.6H2O Formula Weight: 262.86 (154.77anhy) Physical Form: Powder Density: 2.070 Appearance: Blue to bluish green Odour: Odourless pH: 4.3-4.7 Merck Reference: 14,6517 UN No.: 3288 MDL No.: MFCD00149813

Vzorec: NiSO₄·6H₂O
Bod tání: 840 °C
Hustota: 2,070
Storage Temperature: Ambient
MDL Number: MFCD00149813
CAS číslo: 10101-97-0
EINECS: 232-104-9
UN: 3288
ADR: 6.1,III
Merck Index: 14,6517

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR