Tisk…

Lead(II) telluride ≥99.99% (metals basis)

Dodavatel: Thermo Fisher Scientific
Nebezpečí

36317.09 36317.18
36317.09EA 3780 CZK
36317.09 36317.18
Lead(II) telluride ≥99.99% (metals basis)
Lead(II) telluride

Purity: 99.99% CAS No.: 1314-91-6 Molecular Formula: PbTe Formula Weight: 334.79 Physical Form: -325 Mesh Powder Density: 8.164 Melting Point: 917° Merck Reference: 14,5422 UN No.: 3284 MDL No.: MFCD00016274


In semiconductor research, photoconductor cells

Vzorec: PbTe
Bod tání: 917 °C
Hustota: 8,164
Storage Temperature: Ambient
MDL Number: MFCD00016274
CAS číslo: 1314-91-6
EINECS: 215-247-1
UN: 3284
ADR: 6.1,III
Merck Index: 14,5422

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR