Tisk…

Kyselina 4-amino-3-hydroxy-1-naftalensulfonová ≥90% ACS

Dodavatel: Thermo Fisher Scientific
Varování

36290.06 36290.14 36290.22
36290.06EA 1190 CZK
36290.06 36290.14 36290.22
Kyselina 4-amino-3-hydroxy-1-naftalensulfonová ≥90% ACS
Kyselina 4-amino-3-hydroxy-1-naftalensulfonová

Purity: 90% CAS No.: 116-63-2 Molecular Formula: C10H9NO4S Formula Weight: 239.25 Physical Form: Powder Melting Point: ca 295° dec. Beilstein Reference: 2697469 Merck Reference: 14,454 MDL No.: MFCD00004019

Vzorec: C₁₀H₉NO₄S
Bod varu: 466 °C (1013 hPa)
Storage Temperature: Ambient
MDL Number: MFCD00004019
CAS číslo: 116-63-2
EINECS: 204-147-3
Merck Index: 14,454

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR