Tisk…

1-Butanol ≥99.4% ACS, Riedel-de Haën™

Dodavatel: Honeywell Chemicals
Nebezpečí

360465-2.5L 360465-4X2.5L
360465-2.5LEA 4540 CZK
360465-2.5L 360465-4X2.5L
1-Butanol ≥99.4% ACS, Riedel-de Haën™
1-Butanol

ACS GENERAL USE SOLVENTS


  • Value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • Suitable for less demanding applications such as routine isocratic HPLC
  • Variations and decrease issues traceable to solvent contamination
  • All solvents are tested to meet ACS general use specifications


Beilstein Number: 969148
RTECS Number: EO1400000
UNSPSC Code: 12191601

Vzorec: H₃C(CH₂)₂CH₂OH
Bod varu: 119 °C (1,013 hPa)
Bod tání: <-90 °C
Hustota: 0,81 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002964
CAS číslo: 71-36-3
EINECS: 200-751-6
UN: 1120
ADR: 3,III
REACH: 01-2119484630-38
Merck Index: 13,01539
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e9395eeb6150a74222da52460f53cedeccd93442&vid=fd8352a556914c5be7213c576355a88e130d1a7b&prid=web&clid=SAPDAM

Objednat

Výsledky specifikačního testu

Autoignition Temperature 649 °F
Identity (IR) complying
Assay (GC) Min. 99.40 %
Non-volatile matter Max. 0.005 %
Water (Karl Fischer) Max. 0.1 %
Titrable Acid Max. 0.0008 meq/g
Di-n-butyl ether (GC) Max. 0.2 %
n-Butyraldehyde (GC) Max. 0.01 %
APHA Max. 10
Explosion Limit 11.2 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR