Tisk…

Lutécium 1000 mg/L Lu in 5% kyselině dusičné (from Lu₂O₃) Specpure® standard pro ICP

Dodavatel: Thermo Fisher Scientific

35765.AD 35765.AE 35765.AP
35765.ADEA 6010 CZK
35765.AD 35765.AE 35765.AP
Lutécium 1000 mg/L Lu in 5% kyselině dusičné (from Lu₂O₃) Specpure® standard pro ICP
Standardy Standardy kationické Standardy - lutetium

Molecular Formula: Lu₂O3 in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011098


Certifikace: Traceable to NIST

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR