Tisk…

Titanium Standard Solutions for ICP, Specpure®

Dodavatel: Thermo Fisher Scientific

35759.ADEA 2960 CZK
35759.AD 35759.AE 35759.AP 35768.AE 38720.AD 38720.AE 38725.AP 35768.AD 35768.AP 38720.AP 38725.AD 38725.AE 45267.AE
Titanium Standard Solutions for ICP, Specpure®
Standardy Standardy kationické Standardy - titan

UN No.: 3264
MDL No.: MFCD00011264


Certifikace: Traceable to NIST

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR