Tisk…

Jodid zinečnatý, ultra dry ≥99.995% (metals basis)

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: Zinc (II) iodide

Ampuled under argon

35727.06 35727.14 35727.22
35727.06EA 3640 CZK
35727.06 35727.14 35727.22
Jodid zinečnatý, ultra dry ≥99.995% (metals basis)
Jodid zinečnatý

Purity: 99.995% CAS No.: 10139-47-6 Molecular Formula: ZnI2 Formula Weight: 319.18 Physical Form: Granular, Ampouled under argon Density: 4.74 Melting Point: 446° Boiling Point: 625° dec. Merck Reference: 14,10140 UN No.: 3260 MDL No.: MFCD00011299

Vzorec: I₂Zn
Bod varu: 625 °C (rozkl.)
Bod tání: 446 °C
Hustota: 4,74
Storage Temperature: Ambient
MDL Number: MFCD00011299
CAS číslo: 10139-47-6
EINECS: 233-396-0
UN: 3260
ADR: 8,II
Merck Index: 14,10140

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR