Tisk…

Chlorid cínatý, ultra dry ≥99.9985% (metals basis)

Dodavatel: Thermo Fisher Scientific
Nebezpečí


Ampuled under argon

35723.03 35723.06 35723.22 35723.14
35723.03EA 2830 CZK
35723.03 35723.06 35723.22 35723.14
Chlorid cínatý, ultra dry ≥99.9985% (metals basis)
Chlorid cínatý

Purity: 99.9985% CAS No.: 7772-99-8 Molecular Formula: SnCl2 Formula Weight: 189.62 Physical Form: Powder, Ampouled under argon Density: 3.95 Melting Point: 247° Boiling Point: 652° Merck Reference: 14,8783 UN No.: 3260 MDL No.: MFCD00011241

Vzorec: SnCl₂
Bod varu: 652 °C
Bod tání: 247 °C
Hustota: 3,95
Storage Temperature: Ambient
MDL Number: MFCD00011241
CAS číslo: 7772-99-8
EINECS: 231-868-0
UN: 3260
ADR: 8,III
Merck Index: 14,8783

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR