Tisk…

Bromid thalný, ultra dry ≥99.998% (metals basis)

Dodavatel: Thermo Fisher Scientific
Nebezpečí


Ampuled under argon

35720.09 35720.18
35720.09EA 5260 CZK
35720.09 35720.18
Bromid thalný, ultra dry ≥99.998% (metals basis)
Bromid thalný

Purity: 99.998% CAS No.: 7789-40-4 Molecular Formula: TlBr Formula Weight: 284.28 Physical Form: -10 Mesh Beads, Ampouled under argon Density: 7.5 Melting Point: ca 460° Merck Reference: 14,9258 UN No.: 1707 MDL No.: MFCD00011273

Vzorec: BrTl
Bod varu: 815 °C (1013 hPa)
Bod tání: ∼460 °C
Hustota: 7,5
Storage Temperature: Ambient
MDL Number: MFCD00011273
CAS číslo: 7789-40-4
EINECS: 232-163-0
UN: 1707
ADR: 6.1,II
Merck Index: 14,9258

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR