Tisk…

Iron(II) chloride, ultra dry ≥99.99% (metals basis)

Dodavatel: Thermo Fisher Scientific
Nebezpečí


Ampuled under argon

35701.03 35701.09 35701.18
35701.03EA 2580 CZK
35701.03 35701.09 35701.18
Iron(II) chloride, ultra dry ≥99.99% (metals basis)
Iron(II) chloride

Purity: 99.99% CAS No.: 7758-94-3 Molecular Formula: Cl2Fe Formula Weight: 126.75 Physical Form: -10 Mesh Beads, Ampouled under argon Density: 3.160 pH: 2.5 Melting Point: 674° Boiling Point: 1023° Merck Reference: 14,4043 UN No.: 3260 MDL No.: MFCD00011004

Vzorec: Cl₂Fe
Bod varu: 1023 °C
Bod tání: 674 °C
Hustota: 3,160
Storage Temperature: Ambient
MDL Number: MFCD00011004
CAS číslo: 7758-94-3
EINECS: 231-843-4
UN: 3260
ADR: 8,III
Merck Index: 14,4043

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR