Tisk…

Cadmium(II) bromide, ultra dry ≥99.999% (metals basis)

Dodavatel: Thermo Fisher Scientific
Varování


Ampuled under argon

35682.06 35682.14
35682.06EA 6660 CZK
35682.06 35682.14
Cadmium(II) bromide, ultra dry ≥99.999% (metals basis)
Cadmium(II) bromide

Purity: 99.999% CAS No.: 7789-42-6 Molecular Formula: CdBr2 Formula Weight: 272.22 Physical Form: -10 Mesh Beads, Ampouled under argon Density: 5.192 Appearance: White to pale yellow Odour: Odourless Melting Point: 567° Boiling Point: 863° Merck Reference: 14,1615 UN No.: 2570 MDL No.: MFCD00015996

Vzorec: Br₂Cd
Bod varu: 863 °C
Bod tání: 567 °C
Hustota: 5,192
Storage Temperature: Ambient
MDL Number: MFCD00015996
CAS číslo: 7789-42-6
EINECS: 232-165-1
UN: 2570
ADR: 6.1,III
Merck Index: 14,1615

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR