Tisk…

EDTA disodium salt 0.2 M EDTA-Na2 volumetric solution, for complexometry, Fluka™

Dodavatel: Honeywell Chemicals

Synonyms: Idranal III, Edetic acid disodium salt, Kestranal® 2S, Ethylenedinitrilotetra-acetic acid disodium salt, Disodium dihydrogen ethylenediaminetetraacetate, Ethylenediaminetetra-acetic acid disodium salt

35102-5L-VP 35102-1L
35102-5L-VPEA 7630 CZK
35102-5L-VP 35102-1L
EDTA disodium salt 0.2 M EDTA-Na2 volumetric solution, for complexometry, Fluka™
EDTA disodium salt
Vzorec: C₁₀H₁₄N₂Na₂O₈
Storage Temperature: Ambient
MDL Number: MFCD00070672
CAS číslo: 139-33-3
EINECS: 382-266-9

Objednat

Výsledky specifikačního testu

Factor 0.999 - 1.001


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR