Tisk…
Anti-Ly-6G Rat Monoclonal Antibody (FITC (Fluorescein)) [clone: RB6-8C5]
Dodavatel: Tonbo Biosciences
35-5931-U500 35-5931-U100 35-5931-U025
35-5931-U500EA 3370 CZK
35-5931-U500 35-5931-U100 35-5931-U025
Anti-Ly-6G Rat Monoclonal Antibody (FITC (Fluorescein)) [clone: RB6-8C5]
Protilátky
The RB6-8C5 antibody binds to mouse Ly-6G, commonly known as Gr-1, a member of the Ly-6 superfamily of GPI-anchored cell surface proteins with roles in cell signaling and cell adhesion. Gr-1 is differentially expressed during development and maturation of cells in the myeloid lineage and is expression at varying stages and levels on monocytes, macrophages, granulocytes, and peripheral neutrophils.
The RB6-8C5 antibody is typically used in combination with the macrophage labeling antibody M1/70 (Anti-CD11b) for phenotypic analysis of monocytes, macrophages and granulocytes. Note: The RB6-8More Product Information