Tisk…

Mouse IgG2a Isotype Control [clone: C1.18.4] (FITC (Fluorescein Isothiocyanate))

Dodavatel: Tonbo Biosciences
35-4724-U100
35-4724-U100EA 1840 CZK
35-4724-U100
Mouse IgG2a Isotype Control [clone: C1.18.4] (FITC (Fluorescein Isothiocyanate))
Protilátky
The C1.18.4 immunoglobulin is useful as an isotype-matched control. The C.1.18.4 immunolglobulin has an unknown binding specificity and is used as an isotype control for mouse IgG2a antibodies.

Type: Control
Antigen: IgG1 (Mouse) (mouse IgG1)
Clonality: monoclonal
Clone: C1.18.4
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR