Tisk…

n-Pentan ≥99% (by GC), CHROMASOLV™ AMD, Riedel-de Haën™

Dodavatel: Honeywell Chemicals
Nebezpečí

34894-2.5L 34894-4X2.5L
34894-2.5LEA 8540 CZK
34894-2.5L 34894-4X2.5L
n-Pentan ≥99% (by GC), CHROMASOLV™ AMD, Riedel-de Haën™
n-Pentane

  • Solvents designed for high-performance thin-layer chromatography

Vzorec: H₃C(CH₂)₃CH₃
Bod varu: 36-37 °C
Bod tání: -131 °C
Hustota: 0,631 g/cm³ (15 °C)
Bod vzplanutí: <-35 °C
Storage Temperature: Ambient
MDL Number: MFCD00009498
CAS číslo: 109-66-0
EINECS: 203-692-4
UN: 1265
ADR: 3,II
REACH: 01-2119459286-30
Merck Index: 13,07193
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ba2c9c81a4b1f44769627a00ba49b6cc8e63132&vid=f6831eb15ab1602d8c752468dc3ee31cc4ee61c0&prid=web&clid=SAPDAM

Objednat

Výsledky specifikačního testu

Refractive Index n20/D 1.358 (lit.)
Assay (GC) Min. 99 %
Non-volatile matter Max. 0.0005 %
Water (Karl Fischer) Max. 0.005 %
Free acid (as CH₃COOH) Max. 0.0005 %
Transmittance at 200 nm Min. 20 %
Transmittance at 210 nm Min. 60 %
Transmittance at 220 nm Min. 85 %
Transmittance at 230 nm Min. 95 %
Transmittance from 250 nm Min. 99 %
Fluorescence (chinin) at 254 nm Max. 1 ppb
Fluorescence (chinin) at 365 nm Max. 1 ppb
for AMD technic complying
Vapor Density 2.48 (vs air)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR