Tisk…

Methanol ≥99.9%, CHROMASOLV™, gradient grade for HPLC, Riedel-de Haën™

Dodavatel: Honeywell Chemicals
Nebezpečí

34885-7L 34885-18L 34885-45L 34885-1L 34885-2.5L 34885-4X2.5L 34885-6X1L 34885-30L-RC 34885-200L 34885-10L-RC
34885-7L-REA 10790 CZK
34885-7L-R 34885-18L-R 34885-45L-R 34885-1L 34885-2.5L 34885-4X2.5L 34885-6X1L 34885-30L-RC 34885-200L HONC34885-10L-RC
Methanol ≥99.9%, CHROMASOLV™, gradient grade for HPLC, Riedel-de Haën™
Methanol

  • Same high quality as CHROMASOLV HPLC
  • Additional testing for HPLC via gradient analysis and UV-VIS spectroscopy
  • Low UV profile by HPLC
  • Low impurity gradient LC baseline


Beilstein Number: 1098229

Vzorec: CH₃OH
Bod varu: 64…65 °C (1,013 hPa)
Bod tání: –98 °C
Hustota: 0,79 g/cm³ (20 °C)
Bod vzplanutí: 11 °C
Storage Temperature: Ambient
MDL Number: MFCD00004595
CAS číslo: 67-56-1
EINECS: 200-659-6
UN: 1230
ADR: 3,II
REACH: 01-2119433307-44
Merck Index: 13,05984
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ea983428c5015ad8417c1e1c260cce3a4d1de3c&vid=9bc2f21fdce2dcabce477372374ff56b8fb3a0f1&prid=web&clid=SAPDAM

Objednat

Výsledky specifikačního testu

Assay (GC) Min. 99.9 %
Non-volatile matter Max. 0.0003 %
Water (Karl Fischer) Max. 0.02 %
Free alkali (as NH₃) Max. 0.0005 %
Absorbance at 400 nm Max. 0.01
Absorbance at 260 nm Max. 0.01
Absorbance at 240 nm Max. 0.05
Absorbance at 235 nm Max. 0.10
Absorbance at 230 nm Max. 0.15
Absorbance at 220 nm Max. 0.30
Absorbance at 210 nm Max. 0.50
HPLC Gradient at 230 nm Max. 2 mAU
HPLC Gradient at 254 nm Max. 5 mAU
Fluorescence (chinin) at 254 nm Max. 1 ppb
Fluorescence (chinin) at 365 nm Max. 1 ppb
Appearance (color) complying
Appearance (form) complying
Vapor Density 1.11 (vs air)
Vapor Pressure 410 mmHg ( 50 °C)
Autoignition Temperature 725 °F


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR