Tisk…

Ethylacetát ≥99.7%, CHROMASOLV™ for HPLC, Riedel-de Haën™

Dodavatel: Honeywell Chemicals
Nebezpečí

Synonyms: Acetic acid ethyl ester

34858-7L 34858-18L 34858-1L 34858-4X2.5L 34858-6X1L 34858-200L 34858-45L
34858-7LEA 15640 CZK
34858-7L 34858-18L 34858-1L 34858-4X2.5L 34858-6X1L 34858-200L HONC34858-45L
Ethylacetát ≥99.7%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Ethylacetát

  • For HPLC and organic synthesis
  • High-purity, multipurpose solvents
  • High lot-to-lot consistency

Vzorec: CH₃COOC₂H₅
MW: 88.11
Bod varu: 77 °C (1.013 hPa)
Bod tání: –84 °C
Hustota: 0.902
Bod vzplanutí: 11.5 %(V)
Storage Temperature: Ambient
MDL Number: MFCD00009171
CAS číslo: 141-78-6
EINECS: 205-500-4
UN: 1173
ADR: 3 II
REACH: 01-2119475103-46
Merck Index: 13,03792
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=226aecffe42ffab125597552eff0ef7f88ef9983&vid=f5707ce0faee09ca10700419252c5841e0fbc01a&prid=web&clid=SAPDAM

Objednat

Výsledky specifikačního testu

Assay (GC) Min. 99.7 %
Non-volatile matter Max. 0.0005 %
Water (Karl Fischer) Max. 0.03 %
Free acid (as CH₃COOH) Max. 0.005 %
Absorbance at 400 nm Max. 0.01
Absorbance at 300 nm Max. 0.01
Absorbance at 270 nm Max. 0.10
Absorbance at 260 nm Max. 0.30
Absorbance at 255 nm Max. 0.70
Refractive Index n20/D 1.3720 (lit.)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR