Tisk…

Dimethylsulfoxid, extra dry over Molecular Sieve 99.7+%, AcroSeal®

Dodavatel: Thermo Fisher Scientific

Synonyms: Methanesulphinylmethane, Methyl sulphoxide, Dimethyl(oxido)sulphur, DMSO

348440010 348440025 348441000 348445000
348440010.EA 4420 CZK
348440010. 348440025. 348441000. 348445000.
Dimethylsulfoxid, extra dry over Molecular Sieve 99.7+%, AcroSeal®
Dimethylsulfoxid
Vzorec: (CH₃)₂SO
Bod varu: 189 °C (760 mmHg)
Hustota: 1,1 g/cm³
Bod vzplanutí: 87 °C
Storage Temperature: Ambient
MDL Number: MFCD00002089
CAS číslo: 67-68-5
EINECS: 200-664-3
Merck Index: 13,03285
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3ec1573e80c11ddf9254283ef76abf5fb6539c7e&vid=157e2a24c93041cbbb69833d226195e333b07665&prid=web&clid=SAPDAM

Objednat

Výsledky specifikačního testu

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Water ≤0.005 %
GC ≥99.70 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR