Tisk…
HYDRANAL™ humidity absorber for Karl Fischer titration
Dodavatel: Honeywell Chemicals
34788-500G 34788-1KG 34788-6X1KG
34788-500GEA 1600 CZK
34788-500G 34788-1KG 34788-6X1KG
HYDRANAL™ humidity absorber for Karl Fischer titration
Karl Fischer titranty a činidla
HYDRANAL™ Humidity absorber is a drying agent for air and gases for Karl Fischer titration (with indicator).
  • Water absorption capacity: 25%
  • Regeneration: 140 °C (until the colour turns back to orange)