Tisk…

Cyklohexan ≥99.5% (by GC), CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™

Dodavatel: Honeywell Chemicals
Nebezpečí

Synonyms: Hexahydrobenzene, Hexamethylene

34496-7L 34496-18L 34496-1L 34496-2.5L 34496-4X2.5L 34496-7L-RC
34496-7LEA 16470 CZK
34496-7L 34496-18L 34496-1L 34496-2.5L 34496-4X2.5L HONC34496-7L-RC
Cyklohexan ≥99.5% (by GC), CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
Cyklohexan

  • For gas chromatography (GC)
  • Developed especially for the application in residue analysis of pesticides and other low-volatile substances using GC
  • Non-volatile matter and water content are specified for each solvent

Vzorec: C₆H₁₂
Bod varu: 81 °C (1,013 hPa)
Hustota: 0,78 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003814
CAS číslo: 110-82-7
EINECS: 203-806-2
UN: 1145
ADR: 3,II
Merck Index: 13,02752
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8a1e51ba998bef126804c3fd25b361be72cd9545&vid=d1e7f75d9f2a944eb15a7740c5254588cc9a4e67&prid=web&clid=SAPDAM

Objednat

Výsledky specifikačního testu

Assay (GC) Min. 99.5 %
Non-volatile matter Max. 0.001 %
Water (Karl Fischer) Max. 0.01 %
Suitability for residue analysis Max. 5 ng/l
Vapor Density 2.9 (vs air)
Vapor Pressure 168.8 mmHg ( 37.7 °C)
Explosion Limit 9 %
Autoignition Temperature 500 °F


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR