Tisk…

N,N-Dimethylformamid 99,8 GC_ASSAY_METHOD, CHROMASOLV™ Pest_Residual, Riedel-de Haën™

Dodavatel: Honeywell Chemicals
Nebezpečí

Synonyms: Formic acid dimethylamide, Formdimethylamide, Dimethylformamide, DMF

34489-2.5L
34489-2.5LEA 6790 CZK
34489-2.5L
N,N-Dimethylformamid 99,8 GC_ASSAY_METHOD, CHROMASOLV™ Pest_Residual, Riedel-de Haën™
N,N-Dimethylformamid

  • For gas chromatography (GC)
  • Developed especially for the application in residue analysis of pesticides and other low-volatile substances using GC
  • Non-volatile matter and water content are specified for each solvent

Vzorec: HCON(CH₃)₂
MW: 73.09
Bod varu: 152…153 °C (1.013 hPa)
Bod tání: –61 °C
Hustota: 0.94 g/cm³ (20 °C)
Bod vzplanutí: 57.5 °C (1.013.25 hPa)
Storage Temperature: Ambient
MDL Number: MFCD00003284
CAS číslo: 68-12-2
EINECS: 200-679-5
UN: 2265
ADR: 3 III
REACH: 01-2119475605-32
Merck Index: 13,03269
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1ac5c5e9758b6c40ccb0cf127b6a6e1c6d608c64&vid=9a5d4e579e195193602c20d3cb39c89ac9c92cbf&prid=web&clid=SAPDAM

Objednat

Výsledky specifikačního testu

Assay (GC) Min. 99.8 %
Water (Karl Fischer) Max. 0.08 %
Non-volatile matter Max. 0.001 %
Suitability for residue analysis max. 5 ng/l
Explosion Limit 15.2 %
Refractive Index n20/D 1.430 (lit.)
Vapor Pressure 2.7 mmHg ( 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR