Tisk…

thiokyanatan sodný ≥98.0% ACS

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: Sodium rhodanide

33388.36 33388.A3 33388.22
33388.36EA 3220 CZK
33388.36 33388.A3 33388.22
thiokyanatan sodný ≥98.0% ACS
thiokyanatan sodný

Purity: 98.0% min CAS No.: 540-72-7 Molecular Formula: NaSCN Formula Weight: 81.07 Physical Form: Crystalline/Chunks Density: 1.74 Appearance: White Odour: Odourless pH: 6-8 Melting Point: 287° Solubility: Soluble in water, acetone, alcohol Beilstein Reference: 3594965 Merck Reference: 14,9327 MDL No.: MFCD00011123

Vzorec: NaSCN
MW: 81.07
Bod tání: 287
Hustota: 1.735
Storage Temperature: Ambient
MDL Number: MFCD00011123
CAS číslo: 540-72-7
EINECS: 208-754-4
Merck Index: 14,9327

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR