Tisk…
Conical centrifuge tubes with real-time temperature signaling, ViewPoint™
Dodavatel: LABCON
3005-840-000-9 3210-320-008-9 3210-325-008-9 3210-335-008-9 3220-320-008-9 3220-325-008-9 3220-335-008-9
3005-840-000-9EA 11100 CZK
3005-840-000-9 3210-320-008-9 3210-325-008-9 3210-335-008-9 3220-320-008-9 3220-325-008-9 3220-335-008-9
Conical centrifuge tubes with real-time temperature signaling, ViewPoint™
Zkumavky Zkumavky centrifugační
ViewPoint™ tubes are designed to help track the real-time temperature of samples when doing bench work or transferring samples to storage. ViewPoint™ centrifuge tubes visibly signal the temperature of the tube and contents, shifting to dark blue when the temperature of tube contents is below 0 °C.
  • Keep control over work and improve processing times
  • ATP-, RNase/DNase-, and endotoxin-free (non-pyrogenic)
  • Sperm Motility Assay (SMA), Mouse Embryo Assay (MEA) validated safe for cell culture
  • Validated Sterilization (ISO 11137) for use in Research and Medical
More Product Information