Tisk…

Gallium trinitrate hydrate ≥99.9% (metals basis)

Dodavatel: Thermo Fisher Scientific
Nebezpečí

32116.06 32116.14 32116.PG 32116.PF
32116.06EA 2620 CZK
32116.06 32116.14 32116.PG 32116.PF
Gallium trinitrate hydrate ≥99.9% (metals basis)
Gallium trinitrate hydrate

Purity: 99.9% CAS No.: 69365-72-6 Molecular Formula: GaN3O9.xH2O Formula Weight: 255.73(anhy) Physical Form: Lump Sensitivity: Hygroscopic Appearance: White Odour: Odourless Melting Point: 110° dec. Merck Reference: 14,4346 UN No.: 1477 MDL No.: MFCD00149723

Vzorec: GaN₃O₉·H₂O
Storage Temperature: Ambient
MDL Number: MFCD00149723
CAS číslo: 69365-72-6
EINECS: 236-815-5
UN: 1477
ADR: 5.1,III
Merck Index: 14,4346

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR