Tisk…
Ceramic tubes, both ends open, thin wall
Dodavatel: Thermo Fisher Scientific
31706.DR 31706.DM 31708.DM 31708.DR 31708.DW 31709.DM 31709.DR 31709.DW
31706.DREA 2020 CZK
31706.DR 31706.DM 31708.DM 31708.DR 31708.DW 31709.DM 31709.DR 31709.DW
Ceramic tubes, both ends open, thin wall
Hadičky Přenosová hadička
Pure Al₂O₃ Aluminium oxide AL23.
Dense, excellent thermal and electrical resistance properties, corrosion resistant, permeable to microwaves.