Tisk…

Kyselina dusičná 65%, AR®, Macron Fine Chemicals™

Dodavatel: Avantor Performance Materials
Nebezpečí

Synonyms: Nitric acid solution

3131-46EA 12520 CZK
3131-46
Kyselina dusičná 65%, AR®, Macron Fine Chemicals™
Kyselina dusičná
Vzorec: HNO₃
MW: 63.01 g/mol
Bod varu: 87
Bod tání: -42
Hustota: 1.51
MDL Number: MFCD00011349
CAS číslo: 7697-37-2
UN: 2031
ADR: 8 II
Merck Index: 13,06610

Objednat

Výsledky specifikačního testu

Appearance Clear, Colorless Or Yellowish Liquid
Assay ≥65%
Residue after Ignition ≤0.005%
Chloride (Cl) ≤0.0003%
Phosphate (PO₄) ≤0.001%
Sulphate (SO₄) ≤0.001%
Heavy Metals (as Pb) ≤0.0003%
Arsenic (As) ≤0.000003%
Iron (Fe) ≤0.0005%
Lead (Pb) ≤0.0001%
Magnesium (Mg) ≤0.0001%
Manganese (Mn) ≤0.00005%
For Laboratory, Research or Manufacturing Use


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR