Tisk…

1-Decene ≥96% remainder isomers

Dodavatel: Thermo Fisher Scientific
Nebezpečí

31213.22 31213.36 31213.A3
31213.22EA 1120 CZK
31213.22 31213.36 31213.A3
1-Decene ≥96% remainder isomers
1-Decene

Purity: 96% CAS No.: 872-05-9 Molecular Formula: C10H20 Formula Weight: 140.27 Flash Point: 49°(120°F) Physical Form: Liquid Density: 0.741 Refractive Index: 1.4210 Appearance: Colourless Melting Point: -66° Boiling Point: 170-171° Beilstein Reference: 1737236 UN No.: 3295 MDL No.: MFCD00009577

Vzorec: C₁₀H₂₀
Bod varu: 170…171 °C
Bod tání: –66 °C
Hustota: 0,741
Bod vzplanutí: 49 °C (120 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009577
CAS číslo: 872-05-9
EINECS: 212-819-2
UN: 3295
ADR: 3,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR