Tisk…
Bag closures, WeLoc®
Dodavatel: WELOC
S1207001 S1206004 S1207004 S1211001 S1215001 S1222001 S1232001 S1206001_U S1207005 S1205004
312-0106EA 1940 CZK
312-0106 312-0105 312-0107 312-0108 312-0109 312-0110 312-0111 WELOS1206001_U WELOS1207005 WELOS1205004
Bag closures, WeLoc®
Svorky Svorky pro běžné účely
These sealing clips are perfect for use in the demanding pharmaceutical and laboratory environment because they are made from high quality Polyamide 6.6 material from DuPont®. The clips help to maintain hygiene standards, securing opened bags of raw ingredients and materials, such as powders and fluids.
  • Re-closing of partly used bags of ingredients will reduce the risk of cross-contamination
  • Reduce the risk of leakage or accidents
  • Made from strong and durable material
  • Manufactured in Sweden under strict hygiene conditions
  • Clips can be sterilised via
More Product Information