Tisk…

Nessler reagent Ammonia_ammoniasalts_determination

Dodavatel: VWR Chemicals
Nebezpečí

Synonyms: Potassium mercuric Iodide, Potassium tetraiodomercurate, Nota 1, Mercury (II) potassium iodide, Potassium tetraiodomercurate – Potassium hydroxide solution

31074.265EA 1830 CZK
31074.265
Nessler reagent Ammonia_ammoniasalts_determination
Nessler reagent
Bod varu: >100 °C (1013 hPa)
Hustota: 1.2 g/cm³ (20 °C)
MDL Number: MFCD00049656
CAS číslo: 7783-33-7
UN: 2922
ADR: 8 (6.1) II

Objednat

Výsledky specifikačního testu

Sensitivity Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR