Tisk…

Kyselina jodistá 0.1 mol/l (0.1 N) in aqueous solution

Dodavatel: VWR Chemicals

Synonyms: ortho-Periodic acid, Orthoperiodic acid

309746.1000EA 1326 CZK
309746.1000
Kyselina jodistá 0.1 mol/l (0.1 N) in aqueous solution
Kyselina jodistá
Vzorec: H₅IO₆
MW: 227.94
Bod tání: 122
MDL Number: MFCD00011344
CAS číslo: 10450-60-9
EINECS: 233-937-0
Merck Index: 13,07248
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=39e1882cdf619581f467ea2429265a700b5205d5&vid=27e789fa0f30c08d62d5a8d0afc3acd8f6469c12&prid=web&clid=SAPDAM

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR