Tisk…

(±)-1,2-Propandiol ≥99.5% ACS

Dodavatel: Thermo Fisher Scientific

30948.AP 30948.K2 30948.K7 30948.M1
30948.APEA 1810 CZK
30948.AP 30948.K2 30948.K7 30948.M1
(±)-1,2-Propandiol ≥99.5% ACS
(±)-1,2-Propandiol

Purity: 99.5% CAS No.: 57-55-6 Molecular Formula: C3H8O2 Formula Weight: 76.10 Flash Point: 107°(225°F) Physical Form: Liquid Density: 1.036 Refractive Index: 1.4320 Sensitivity: Hygroscopic Odour: Nearly Odourless pH: neutral Melting Point: -60° Boiling Point: 186-188° Solubility: Miscible with water, acetone, chloroform. Soluble in ether Beilstein Reference: 1340498 Merck Reference: 14,7855 MDL No.: MFCD00064272


Common Applications: As a substitute for ethylene glycol and glycerol. Good solvent for resins. Dissolves many essential oils, but immiscible with fixed oils.

Vzorec: CH₃CH(OH)CH₂OH
Bod varu: 186…188 °C
Bod tání: –60 °C
Hustota: 1,036
Bod vzplanutí: 107 °C (225 °F)
Storage Temperature: Ambient
MDL Number: MFCD00064272
CAS číslo: 57-55-6
EINECS: 200-338-0
Merck Index: 14,7855
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f2af195b87767feb8d1baca34925153d6f97ae2d&vid=a598a9042847dc3ce52daed1a9326b592b163170&prid=web&clid=SAPDAM

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR