Tisk…

Tetrasodium meso-tetra(sulphonatophenyl)porphine dodecahydrate ≥95%

Dodavatel: Thermo Fisher Scientific
Varování

30538.03 30538.MD
30538.03EA 10570 CZK
30538.03 30538.MD
Tetrasodium meso-tetra(sulphonatophenyl)porphine dodecahydrate ≥95%
Tetrasodium meso-tetra(sulphonatophenyl)porphine dodecahydrate

Purity: 95% CAS No.: 122451-08-5 Molecular Formula: C44H26N4Na4O12S4.12H2O Formula Weight: 1239.11 (1022.87anhy) Physical Form: Crystalline Beilstein Reference: 5488862 MDL No.: MFCD00151011

Vzorec: C₄₄H₂₆N₄Na₄O₁₂S₄·12H₂O
Storage Temperature: Ambient
MDL Number: MFCD00151011
CAS číslo: 122451-08-5
EINECS: 254-262-8

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR