Tisk…

Hydrogenfosforečnan dvojsodný dihydrát 98.5-101.0% (dried basis), Puriss. p.a. Reag. Ph. Eur., Fluka™

Dodavatel: Honeywell Chemicals

Synonyms: sec-Sodium phosphate, Sodium monohydrogen phosphate dihydrate, Sodium hydrogenphosphate dihydrate, di-Sodium hydrogen orthophosphate dihydrate, Sodium phosphate dibasic dihydrate, Disodium phosphate, di-Sodium hydrogen phosphate dihydrate, Sorensen's salt

30435-1KG 30435-500G 30435-6X1KG 30435-5KG
30435-1KGEA 2090 CZK
30435-1KG 30435-500G 30435-6X1KG 30435-5KG
Hydrogenfosforečnan dvojsodný dihydrát 98.5-101.0% (dried basis), Puriss. p.a. Reag. Ph. Eur., Fluka™
Hydrogenfosforečnan dvojsodný dihydrát
Vzorec: Na₂HPO₄·2H₂O
MW: 177.99
Bod tání: 450 °C
Hustota: 2.1
Storage Temperature: Ambient
MDL Number: MFCD00149182
CAS číslo: 10028-24-7
EINECS: 231-448-7

Objednat

Výsledky specifikačního testu

Assay (calculated to the dried substance) 98.5 - 101.0 %
Assay of NaH₂PO₄ (calculated to dried substances) Max. 0.5 %
Loss on drying (130°C) 19.5 - 21.0 %
pH (5 %, 20°C) 9.0 - 9.2
Arsenic (As) Max. 0.00005 %
Iron (Fe) Max. 0.0005 %
Potassium (K) Max. 0.005 %
Heavy metals (as Pb) Max. 0.0005 %
Chloride (Cl) Max. 0.001 %
Sulfate (SO₄) Max. 0.005 %
Total N Max. 0.001 %
Appearance of the solution complying
Reducing impurities complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR