Tisk…
Cell surface staining kits, MemBrite™ Fix
Dodavatel: Biotium
30092-T 30092 30093-T 30094 30094-T 30097-T 30093 30095-T 30095 30096-T 30096 30097 30098-T 30098 30099-T 30099 30103-T 30101-T 30101 30102-T 30102 30103 30104-T 30104
30092-TEA 8320 CZK
30092-T 30092. 30093-T 30094. 30094-T 30097-T 30093. 30095-T 30095. 30096-T 30096. 30097. 30098-T 30098. 30099-T 30099. 30103-T 30101-T 30101. 30102-T 30102. 30103. 30104-T 30104.
Cell surface staining kits, MemBrite™ Fix
Sítka pro tkáňové kultury
MemBrite™ Fix cell surface staining kits are designed for covalently staining the surface of live cells to conveniently visualise cell boundaries in immunofluorescence experiments.
  • Rapid, uniform live cell surface staining
  • Covalent fluorescent labelling can be fixed and permeabilised for IF staining
  • Stain mammalian cells, yeast, or gram-positive bacteria
  • Choose from 12 bright and photostable dye colours from blue to near-IR
  • MemBrite™ Fix-ST dyes for super-resolution STORM imaging
These stains are novel reactive fluorescent dyes stains that reactMore Product Information