Tisk…
Dipping bottle, 'Ex'
Dodavatel: BURKLE
5365-3000
300-1035EA 17810 CZK
300-1035
Dipping bottle, 'Ex'
Vzorkovače Naběračky na vzorky
Dipping bottle for testing mineral oil products (Category A, Explosion Categories IIA, IIB and IIC), crude oil, liquid mineral oil products, fuels, special and test fuels, fuel oils, petroleum and liquid lubricants. Completely made from non spark-producing materials, nickel plated.
  • Screw-off bottle head
  • Easy to clean robust handle
  • High weight (3,2 kg) for fast lowering
Certifikace: Tested for use in the Ex protection zone 0.
Informace pro objednávání: Please order the appropriate lowering cable at the same time.