Tisk…
Drums, stainless steel
Dodavatel: Sampling Systems
A196-05 A196-10 A196-100 A196-15 A196-150 A196-20 A196-200 A196-25 A196-30 A196-50 A196-75
SAMPA196-05EA 22650 CZK
SAMPA196-05 SAMPA196-10 300-0627 300-0628 300-0629 300-0630 300-0631 300-0632 300-0633 300-0634 300-0635
Drums, stainless steel
Sudy, soudky, bubny
These stainless steel containers are made from high-quality 316L stainless steel and are ideal for almost all aplications, including GMP use.
  • Suitable for pharmaceutical, chemical, food, and cosmetic manufacturing
  • Complete with lid, clamp, and FDA compliant seal
  • Extremely easy and secure closing and reopening
  • High-quality mirror finish
  • Fast delivery
This range of stainless steel containers is perfect for dispensing liquids. Due to their ease of use, they are very popular across many different industries.
Informace pro objednávání: Emptying tap hasMore Product Information